Learn More
Human CD11c Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB17771-SP
This item is not returnable.
View return policy
Description
CD11c Monoclonal specifically detects CD11c in Human samples. It is validated for Western Blot.
Specifications
| CD11c | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 0.5-15 ug/mL | |
| P20702 | |
| ITGAX | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD11c aa 20-1107 Accession # P20702 | |
| 25 μg | |
| Primary | |
| Detects human CD11c in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Integrin alpha 1, 2, 4, 5, 6, 8, 9, 10, 11, L, M, rhIntegrin alpha 2b/beta 3, alpha 3/beta 1, alpha 6X1/beta 4, alpha 7X2/beta 1, alpha D/beta 2, alpha E/beta 7, alpha V/beta 6, recombinant mouse Integrin alpha X, and alpha 7X2 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 538207 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, Integrin alpha X, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), ITGAX, Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, p150,95 alpha, SLEB6 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3687 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |