Learn More
Human Cornulin Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3607-SP
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Cornulin Polyclonal specifically detects Cornulin in Human samples. It is validated for Western Blot.
Tekniske data
| Cornulin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 53 kDa squamous epithelial-induced stress protein, 58 kDa heat shock protein, C1orf10, chromosome 1 open reading frame 10, cornulin, CRNN, DRC1, Gm1015, PDRC1, PDRC1,53 kDa putative calcium-binding protein, SEP53, SEP53C1orf10, Squamous epithelial heat shock protein 53, Tumor-related protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 49860 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q9UBG3 | |
| CRNN | |
| E. coli-derived recombinant human Cornulin Met1-Ser140 Accession # Q9UBG3 | |
| 25 μg | |
| Primary | |
| Detects human Cornulin in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion