Learn More
Rad51C Antibody (2H11/6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 29 publications
Brand: Novus Biologicals NB100-177
This item is not returnable.
View return policy
Description
Rad51C Monoclonal specifically detects Rad51C in Human, Mouse, Primate, Yeast samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| Rad51C | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Simple Western 1:100, Flow Cytometry 1 ug per million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin reported in scientific literature (PMID 23512992), CyTOF-ready | |
| BROVCA3, DNA repair protein RAD51 homolog 3, FANCO, MGC104277, R51H3, RAD51 homolog C, RAD51 homolog C (S. cerevisiae), RAD51L2RAD51 (S. cerevisiae) homolog C, RAD51-like protein 2, yeast RAD51 homolog 3 | |
| Mouse | |
| 40 kDa | |
| 0.1 mL | |
| Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination | |
| 5889 | |
| Human, Mouse, Primate, Yeast | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 2H11/6 | |
| Unconjugated | |
| O43502 | |
| RAD51C | |
| His-tagged human Rad51C, over-expressed in E. coli. [UniProt# O43502] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Does not cross-react with Rad51B, Rad51D, Rad51, XRCC2, or XRCC3 in Western analysis. | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |