Learn More
Human/Mouse/Rat PKLR Antibody, R&D Systems™
Rabbit Polyclonal Antibody
Brand: R&D Systems AF8519-SP
This item is not returnable.
View return policy
Description
PKLR Polyclonal specifically detects PKLR in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PKLR | |
| Polyclonal | |
| Western Blot 1:1000 dilution, Simple Western 0.5 μg/mL | |
| P30613 | |
| PKLR | |
| E. coli-derived recombinant human PKLR Leu47-Ser574 Accession # P30613 | |
| 25 μL | |
| Primary | |
| Detects human, mouse, and rat PKLR in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human PKM-2 is observed. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. |
| Western Blot | |
| Unconjugated | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| EC 2.7.1.40, PK1, PK1PKRL, PKL, PKR, Pyruvate kinase 1, pyruvate kinase isozyme R/L, pyruvate kinase isozymes R/L, pyruvate kinase type L, pyruvate kinase, liver and blood cell, pyruvate kinase, liver and RBC, Red cell/liver pyruvate kinase, RPK, R-type/L-type pyruvate kinase | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 5313 | |
| Human, Mouse, Rat | |
| IgG |