Learn More
Human IFN-alpha/beta R2 C-Terminus Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF7014-SP
This item is not returnable.
View return policy
Description
IFN-alpha/beta R2 Polyclonal specifically detects IFN-alpha/beta R2 in Human samples. It is validated for Western Blot.
Specifications
| IFN-alpha/beta R2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta receptor10, human interferon alpha, human interferon alpha/beta receptor, IFNABR, IFN-alpha binding protein, IFN-alpha/beta receptor 2, IFN-alpha-REC, IFN-aR2, IFNAR2, IFNARB, IFNbR2, IFN-bR2, IFN-R, IFN-R-2, interferon (alpha, beta and omega) receptor 2, Interferon alpha binding protein, interferon alpha/beta receptor 2, interferon-alpha/beta receptor beta chain, Type I interferon receptor 2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3455 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P48551 | |
| IFNAR2 | |
| E. coli-derived recombinant human IFN-alpha/beta R2 C-Terminus Ser351-Met514 Accession # P48551 | |
| 25 μg | |
| Primary | |
| Detects human IFN-alpha/beta R2 C-Terminus in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) IFN-alpha/beta R2a, rhIFN-alpha/beta R alpha , and recombinant mouse IFN-alpha/beta R2 is observed. | |
| Human | |
| IgG |