Learn More
Human/Primate CXCL1/GRO alpha/KC/CINC-1 Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems IC2751U-100UG
Product Code. 18772768
  • £461.00 / 100µg
Description
CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Human, Primate samples. It is validated for Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCL1/GRO alpha/KC/CINC-1
Monoclonal
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Mouse
100 μg
Primary
Detects human and primate CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Flow Cytometry
20326R
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1
E. coli-derived recombinant human CXCL1/GRO alpha/KC/CINC-1
RUO
2919
Human, Primate
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only (RUO)