Learn More
Mouse/Rat Tenascin R Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB1624-SP
This item is not returnable.
View return policy
Description
Tenascin R Monoclonal specifically detects Tenascin R in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Tenascin R | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Janusin, MGC149328, Restrictin, tenascin R (restrictin, janusin), tenascin-R, TNR, TN-R | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7143 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 619 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TNR | |
| ôrest L2ö fraction from bovine brain | |
| 25 μg | |
| Primary | |
| Detects mouse and rat Tenascin R in Western blots. | |
| Mouse, Rat | |
| Purified |