Learn More
Novus Biologicals™ ACBP Protein
Product Code. 18766213 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.5mg
Product Code. 18766213
Brand: Novus Biologicals™ NBP1508330.5mg
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-87 of Human ACBP The Recombinant Human ACBP Protein is derived from E. coli. The Recombinant Human ACBP Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.5 mg