Learn More
Human/Rat Annexin V Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB399-100
This item is not returnable.
View return policy
Description
Annexin V Monoclonal specifically detects Annexin V in Human, Rat samples. It is validated for Western Blot, Simple Western, Knockout Validated.
Specifications
| Annexin V | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, ANXA5, ANXV, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha | |
| E. coli-derived recombinant human Annexin V Met1-Asp320 Accession # P08758 | |
| 100 μg | |
| Primary | |
| Detects human Annexin V in direct ELISAs and Western blots. Detects rat Annexin V in Simple Westerns TM . | |
| Human, Rat | |
| Purified |
| Western Blot | |
| 39103 | |
| Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL | |
| P08758 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 308 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction