Learn More
Human/Mouse/Rat MCHR1 Antibody, R&D Systems™
Description
MCHR1 Monoclonal specifically detects MCHR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.
Specifications
Specifications
| Antigen | MCHR1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 851047 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Simple Western 50 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | Q99705 |
| Gene Alias | G protein-coupled receptor 24, GPR24, GPR24SLC-1, G-protein coupled receptor 24, MCH receptor 1, MCH1R, MCH-1R, MCH1RMCHR-1, MCH-R1, melanin-concentrating hormone receptor 1, MGC32129, SLC1, SLC1MCHR, Somatostatin receptor-like protein |
| Show More |