Learn More
Human Neurotrimin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1235-SP
This item is not returnable.
View return policy
Description
Neurotrimin/HNT Monoclonal specifically detects Neurotrimin/HNT in Human samples. It is validated for Western Blot.
Specifications
| Neurotrimin/HNT | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9P121 | |
| NTM | |
| Mouse myeloma cell line NS0-derived recombinant human Neurotrimin Val31-Asn321 Accession # Q9P121 | |
| 25 μg | |
| Primary | |
| Detects human Neurotrimin in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 175214 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hNT, IgLON family member 2, IGLON2HNTNT, MGC60329, neurotrimin, NTRI | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 50863 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |