Learn More
Human/Mouse STAT5a Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2174-SP
This item is not returnable.
View return policy
Description
STAT5A Monoclonal specifically detects STAT5A in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| STAT5a | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| NP_003143 | |
| STAT5A | |
| Human STAT5a synthetic peptide SLDSRLSPPAGLFTSARGSLS Accession # NP_003143 | |
| 25 μg | |
| Primary | |
| Detects human and mouse STAT5a. Does not cross-react with STAT5b. | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 251619 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGF, signal transducer and activator of transcription 5A, STAT5 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6776 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG3 |