Learn More
Human BCR Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF5129-SP
This item is not returnable.
View return policy
Description
BCR Polyclonal specifically detects BCR in Human samples. It is validated for Western Blot.
Specifications
| BCR | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ALL, ALLD22S11, BCR1, breakpoint cluster region, breakpoint cluster region protein, CMLEC 2.7.11.1, D22S662, PHL, PHLFLJ16453, Renal carcinoma antigen NY-REN-26 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 613 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P11274 | |
| BCR | |
| E. coli-derived recombinant human BCR Lys174-Asp331 Accession # P11274 | |
| 25 μg | |
| Primary | |
| Detects human BCR in Western blots. | |
| Human | |
| IgG |