Learn More
Mouse VLDLR Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB2258-SP
This item is not returnable.
View return policy
Description
VLDLR Monoclonal specifically detects VLDLR in Mouse samples. It is validated for Western Blot.
Specifications
| VLDLR | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P98156 | |
| VLDLR | |
| Mouse myeloma cell line NS0-derived recombinant mouse VLDL R Thr25-Ala798 Accession # P98156 | |
| 25 μg | |
| Primary | |
| Detects mouse VLDL R in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human ApoER2. | |
| Mouse | |
| Purified |
| Western Blot | |
| 268724 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CARMQ1, CHRMQ1, FLJ35024, very low density lipoprotein receptor, very low-density lipoprotein receptor, VLDL R, VLDL receptor, VLDL-R, VLDLRCH | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7436 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |