Learn More
R&D Systems™ Recombinant Human Slit3 (aa 1120-1523) Protein
Product Code. 18717979
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. 18717979
Brand: R&D Systems™ 9067SL050
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
The Recombinant Human Slit3 (aa 1120-1523) Protein is derived from HEK293. The Recombinant Human Slit3 (aa 1120-1523) Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number O75094
For Use With (Application) Bioactivity
Gene ID (Entrez) 6586
Molecular Weight (g/mol) Observed M.W.: 56-63kDa, reducing conditions, Theoretical M.W.: 45kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human Slit3 protein Thr1120-Ser1523,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias KIAA0814, MEGF5, MEGF5Multiple epidermal growth factor-like domains protein 5, Multiple EGF-like domains protein 5, SLIL2, SLIL2FLJ10764, slit homolog 3 (Drosophila), slit homolog 3 protein, SLIT1, slit2, Slit-3slit (Drosophila) homolog 3
Common Name Slit3
Gene Symbol SLIT3
Biological Activity Measured by its ability to enhance neurite outgrowth of E16-E18 rat embryonic cortical neurons. Recombinant Human Slit3, immobilized at 1.25-2.5μg/mL on a 96-well plate, is able to significantly enhance neurite outgrowth.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Thr1120-Ser1523, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining