Learn More
Human IgM Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9435-SP
This item is not returnable.
View return policy
Description
IgM Monoclonal specifically detects IgM in Human samples. It is validated for Western Blot, Simple Western, ELISA.
Specifications
| IgM | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3507 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, ELISA | |
| 2123A | |
| Western Blot 0.1 ug/mL, Simple Western 5 ug/mL, ELISA | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Immunoglobulin M (IgM), mu chain from human plasma | |
| 25 μg | |
| Primary | |
| Detects human IgM in Western blots. | |
| Human | |
| Purified |
Product Content Correction
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction