Learn More
Human/Mouse/Rat Axin-1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 7 publications
Brand: R&D Systems AF3287-SP
This item is not returnable.
View return policy
Description
Axin-1 Polyclonal specifically detects Axin-1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Axin-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| axin 1, axin-1, Axin1, AXINaxis inhibitor 1, Axis inhibition protein 1, fused, mouse, homolog of, hAxin, MGC52315 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8312 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| O15169 | |
| AXIN1 | |
| E. coli-derived recombinant human Axin-1 Tyr210-Glu410 Accession # O15169 | |
| 25 μg | |
| Primary | |
| Detects human, mouse and rat Axin-1 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |