Learn More
R&DSystems™ Recombinant Human Sialate O-acetylesterase/SIAE His-tag, CF
Click to view available options
:
20 μg
Description
Measured by its ability to hydrolyze p-nitrophenylacetate. The specific activity is >25000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | Q9HAT2 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Molecular Weight (g/mol) | M.W.-observed: 63-69 & 88-97 kD, under reducing conditions., M.W.-theroretical: 57 kD |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human SIAE protein Ile24-Lys523, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | AIS6, Bakers yeast, Baker's yeast, Bakers' yeast, Brewers yeast, Brewer's yeast, Brewers' yeast, CSEC, CSE-C, EC 3.1.1.53, LSE, MGC87009, S cerevisiae, S. cerevisiae, S.cerevisiae, Sialate O-Acetylesterase, YSG2 |
| Conjugate | Unconjugated |
| Show More |