Learn More
Human/Rat Neuroligin 2/NLGN2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF5645-SP
This item is not returnable.
View return policy
Description
Neuroligin 2/NLGN2 Polyclonal specifically detects Neuroligin 2/NLGN2 in Human, Rat samples. It is validated for Western Blot.
Specifications
| Neuroligin 2/NLGN2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.1.1, EC 3.1.1.1, KIAA1366neuroligin-2, neuroligin 2, NLGN2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 57555 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1-3 ug/mL | |
| EAW90197 | |
| NLGN2 | |
| Mouse myeloma cell line NS0-derived recombinant human Neuroligin 2/NLGN2 Gln15-Ser660 Accession # EAW90197 | |
| 25 μg | |
| Primary | |
| Detects human and rat Neuroligin 2/NLGN2 in direct ELISAs and Western blots. | |
| Human, Rat | |
| IgG |