Learn More
Mouse Neutrophil Elastase/ELA2 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF4517-SP
This item is not returnable.
View return policy
Description
Neutrophil Elastase/ELA2 Polyclonal specifically detects Neutrophil Elastase/ELA2 in Mouse samples. It is validated for Western Blot.
Specifications
| Neutrophil Elastase/ELA2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2, ELA2granulocyte-derived elastase, ELANE, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, Leukocyte Elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1991 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q3UP87 | |
| ELANE | |
| Mouse myeloma cell line NS0-derived recombinant mouse Neutrophil Elastase/ELA2 Ser27-Arg260 Accession # Q3UP87 | |
| 25 μg | |
| Primary | |
| Detects mouse Neutrophil Elastase/ELA2 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human ELA2 is observed. | |
| Mouse | |
| IgG |