Learn More
Human/Mouse/Rat GULP1/CED-6 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF7978-SP
This item is not returnable.
View return policy
Beskrivning
GULP1/CED-6 Polyclonal specifically detects GULP1/CED-6 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifikationer
| GULP1/CED-6 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CED6, CED6GULPCED-6, Cell death protein 6 homolog, engulfment adapter protein, FLJ31156, GULP, engulfment adaptor PTB domain containing 1, Protein GULP, PTB domain adapter protein CED-6, PTB domain adaptor protein CED-6, PTB domain-containing engulfment adapter protein 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 51454 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9UBP9 | |
| GULP1 | |
| E. coli-derived recombinant human GULP1/CED-6 Met1-Asn92 Accession # Q9UBP9 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat GULP1/CED-6 in direct ELISAs and Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering