Learn More
Human/Mouse/Rat AIF Biotinylated Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  R&D Systems BAF1457
Product Code. 18707977
  • £457.00 / 50µg
Description
AIF Polyclonal specifically detects AIF in Human, Mouse, Rat samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AIF
Polyclonal
Biotin
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
AIFapoptosis-inducing factor 1, mitochondrial, AIFM1, apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, MGC111425, PDCD8, PDCD8striatal apoptosis-inducing factor, programmed cell death 8 (apoptosis-inducing factor), Programmed cell death protein 8
Rabbit
Antigen Affinity-purified
Primary
AIF antibodies are ideal for immunocytochemistry colocalization studies in mitochondria. The unconjugated antibody detects human, mouse, and rat mitochondria-processed AIF.
Human, Mouse, Rat
Purified
Immunocytochemistry
LYOPH
Immunocytochemistry 5-15 ug/mL
O95831
AIFM1
E. coli-derived recombinant human AIF Glu121-Asp613 Accession # O95831
50 μg
9131
Reconstitute at 0.2 mg/mL in sterile PBS.