Learn More
Mouse XCL1/Lymphotactin Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB486-SP
This item is not returnable.
View return policy
Description
XCL1/Lymphotactin Monoclonal specifically detects XCL1/Lymphotactin in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| XCL1/Lymphotactin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ATAC, ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1 alpha, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin) | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6375 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, ELISA | |
| 80222 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| AAA56752 | |
| XCL1 | |
| E. coli-derived recombinant mouse XCL1/Lymphotactin Val22-Gly114 Accession # AAA56752 | |
| 25 μg | |
| Primary | |
| Detects mouse XCL1/Lymphotactin in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant human XCL1/Lymphotactin is observed and no cross-reactivity with recombinant viral (rv) CMV UL147 is observed. In Western blots, 75% cross-reactivity with rvCMV UL147 is observed and no cross-reactivity with recombinant human CCL1-3, 5, 7, 8, 11, 13-25, 28, recombinant mouse CCL1-7, 9, 11, 12, 15, 19, 21, 22, 24, 25, 27 or 28 is observed. | |
| Mouse | |
| Purified |