Learn More
Tocris Bioscience™ LW 6
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
LW 6 is a malate dehydrogenase-2 (MDH2) inhibitor (IC50 = 6.3 μM). Also inhibits HIF-1α. Prevents accumulation of HIF-1α in Hep3B cells under hypoxic conditions (IC50 = 2.6 μM) with resulting decrease in expression of HIF-1α target genes VEGF and EPO. Inhibits proliferation of activated human T cells ex vivo.
Specifications
Specifications
| Chemical Name or Material | Methyl 4-hydroxy-3-[[2-(4-tricyclo[3.3.1.13,7 ]dec-1-ylphenoxy)acetyl]amino]benzoate |
| CAS | 934593-90-5 |
| Quantity | 10 mg |
| Target | Dehydrogenase Inhibitors |
| Molecular Formula | C26H29NO5 |
| Purity | 0.98 |