Learn More
Human GRK5 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB4539
Product Code. 18703677
  • £346.00 / 100µg
Description
GRK5 Monoclonal specifically detects GRK5 in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GRK5
Monoclonal
LYOPH
Immunocytochemistry 8-25 ug/mL
EC 2.7.11, EC 2.7.11.16, FLJ39780, G protein-coupled receptor kinase 5, G protein-coupled receptor kinase GRK5, GPRK5, GPRK5FP2025
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human GRK5 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) GRK6 is observed and no cross-reactivity with rhGRK4 is observed.
Human
Purified
Immunocytochemistry
691816
Unconjugated
P34947
GRK5
E. coli-derived recombinant human GRK5 Pro463-Ser590 Accession # P34947
100 μg
2869
Sterile PBS to a final concentration of 0.5 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a