Learn More
Human/Mouse/Rat FKBP12 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB3777-SP
This item is not returnable.
View return policy
Description
FKBP12 Monoclonal specifically detects FKBP12 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| FKBP12 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P62942 | |
| FKBP1A | |
| E. coli-derived recombinant human FKBP12 Gly2-Glu108 Accession # P62942 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat FKBP12 in Western blots. In Western blots, no cross-reactivity with other FKBP family members is observed. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 422513 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP1a, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PKC12, PPIASE, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2280 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |