Learn More
Human BMP-4 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 4 publications
Brand:  R&D Systems AF757-SP
Product Code. 18700299
  • £170.00 / 25µg
Description
BMP-4 Polyclonal specifically detects BMP-4 in Human samples. It is validated for Western Blot, Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BMP-4
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME
Goat
Affinity Purified
RUO
652
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Neutralization
LYOPH
Western Blot 0.1 ug/mL, Neutralization 0.5-2.0 ug/mL
P12644
BMP4
Mouse myeloma cell line NS0-derived recombinant human BMP-4 Ser293-Arg408 Accession # P12644
25 μg
Primary
Detects human BMP-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human (rh) BMP-2 and 10% cross-reactivity with rhBMP-5 and rhBMP-6 is observed.