Learn More
Contactin-2/TAG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99313-50ul
This item is not returnable.
View return policy
Description
Contactin-2/TAG1 Polyclonal antibody specifically detects Contactin-2/TAG1 in Human samples. It is validated for ELISA
Specifications
| Contactin-2/TAG1 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| Axonal glycoprotein TAG-1, Axonin-1, axonin-1 cell adhesion molecule, AXT, contactin 2 (axonal), contactin 2 (transiently expressed), contactin-2, DKFZp781D102, FLJ37193, MGC157722, TAG1, TAG-1, TAX, TAX-1, TAX1FLJ42746, Transient axonal glycoprotein 1, transiently-expressed axonal glycoprotein | |
| Produced in rabbits immunized with purified, recombinant Human Contactin-2/TAG1 (Accession#: NP_005067.1; Met1-Asn1012) | |
| 50 μg | |
| Cancer, Cell Biology, Immunology, Neuroscience | |
| 6900 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |