Learn More
GCET2 Antibody (HGAL/2373), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-14265APC
Product Code. 18669138
  • £532.00 / 0.10mL
Description
GCET2 Monoclonal antibody specifically detects GCET2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GCET2
Monoclonal
APC
GAL, GCAT2, germinal center B-cell-expressed transcript 2 protein, germinal center expressed transcript 2, Germinal center-associated lymphoma protein, HGAL, MGC40441
Recombinant fragment (178 Amino acid residues from around aa1-200) of human GCET2 protein (Uniprot: Q8N6F7)
0.1 mL
Cancer, Immunology
257144
Store at 4°C in the dark.
IgG2b κ
Immunohistochemistry (Paraffin), Protein Array
HGAL/2373
PBS
Mouse
Protein A or G purified