Description
PEPT1/SLC15A1 Polyclonal antibody specifically detects PEPT1/SLC15A1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | PEPT1/SLC15A1 |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Formulation | PBS |