Learn More
COPS7A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92685-0.1ml
This item is not returnable.
View return policy
Description
COPS7A Polyclonal antibody specifically detects COPS7A in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| COPS7A | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| COP9 (constitutive photomorphogenic, Arabidopsis, homolog) subunit 7A, COP9 complex subunit 7a, COP9 constitutive photomorphogenic homolog subunit 7A (Arabidopsis), COP9 signalosome complex subunit 7a, CSN7AMGC110877, Dermal papilla-derived protein 10, DERP10, JAB1-containing signalosome subunit 7a, SGN7a, Signalosome subunit 7a | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-275 of human COPS7A (NP_057403.1). MSAEVKVTGQNQEQFLLLAKSAKGAALATLIHQVLEAPGVYVFGELLDMPNVRELAESDFASTFRLLTVFAYGTYADYLAEARNLPPLTEAQKNKLRHLSVVTLAAKVKCIPYAVLLEALALRNVRQLEDLVIEAVYADVLRGSLDQRNQRLEVDYSIGRDIQRQDLSAIARTLQEWCVGCEVVLSGIEEQVSRANQHKEQQLGLKQQIESEVANLKKTIKVTTAAAAAATSQDPEQHLTELREPAPGTNQRQPSKKASKGKGLRGSAKIWSKSN | |
| 0.1 mL | |
| Cell Biology | |
| 50813 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |