Learn More
CRISP-3 Antibody (295208), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB23972A
Product Code. 18648443
  • £463.00 / 0.10mL
Description
CRISP-3 Monoclonal antibody specifically detects CRISP-3 in Human samples. It is validated for Flow Cytometry (Intracellular Staining)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CRISP-3
Monoclonal
APC
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
10321
Store at 4°C in the dark.
IgG2b
Flow Cytometry (Intracellular Staining)
295208
Intracellular Staining by Flow Cytometry
CRISP-3MGC126588, CRS3, cysteine-rich secretory protein 3, cysteine-rich secretory protein-3, dJ442L6.3, SGP28Aeg2, specific granule protein (28 kDa), Specific granule protein of 28 kDa
Mouse myeloma cell line NS0-derived recombinant human CRISP-3, Asn21-Tyr245 (Ser106Pro&Ala134Ser), Accession # P54108.1
0.1 mL