Learn More
UFD1L Antibody (S09-7G5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19979-50ul
This item is not returnable.
View return policy
Description
UFD1L Monoclonal antibody specifically detects UFD1L in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| UFD1L | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7353 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| S09-7G5 | |
| Unconjugated | |
| ATP1C, ATP1G1ATPase, Na+/K+ transporting, gamma 1 polypeptide, FXYD domain containing ion transport regulator 2, FXYD domain-containing ion transport regulator 2, HOMG2, hypomagnesemia 2, renal, MGC12372, Na(+)/K(+) ATPase subunit gamma, Sodium pump gamma chain, sodium/potassium-transporting ATPase subunit gamma, Sodium-potassium-ATPase, gamma polypeptide | |
| A synthetic peptide of human UFD1LL (Uniprot # Q92890) | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |