Learn More
Novus Biologicals™ RPP38 Recombinant Protein Antigen
Product Code. 18636717 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 18636717
Brand: Novus Biologicals™ NBP268691PEP
Recombinant Protein
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Antibody Competition
Formulation PBS and 1 M Urea, pH 7.4.
Gene ID (Entrez) 10557
Molecular Weight (g/mol) MolecularWeight-theroretical: 27 kDa
Quantity 100 μL
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Gene Alias EC 3.1.26.5, ribonuclease P (38 kD) (RPP38), ribonuclease P protein subunit p38, ribonuclease P/MRP 38 kDa subunit, RNaseP protein p38
Gene Symbol RPP38
Inhibitor Properties This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-68691. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL
Research Category Endocrinology
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein RPP38