Learn More
DRAP1 Antibody (PCRP-DRAP1-1A12), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24152H
Product Code. 18635004
  • £514.00 / 0.10mL
Description
DRAP1 Monoclonal antibody specifically detects DRAP1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DRAP1
Monoclonal
HRP
dr1-associated corepressor, Dr1-associated protein 1, DR1-associated protein 1 (negative cofactor 2 alpha), NC2-alphaNegative co-factor 2-alpha, negative cofactor 2 alpha
Recombinant human DRAP1 protein
0.1 mL
Primary
Human
Purified
Flow Cytometry, Immunofluorescence
PCRP-DRAP1-1A12
PBS
Mouse
Protein A or G purified
RUO
10589
Store at 4°C in the dark.
IgG2b κ