Learn More
Apolipoprotein B/ApoB Antibody (SR1898), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21714-100ul
 View more versions of this product
Product Code. 18632845
  • £341.00 / 100µL
Description
Apolipoprotein B/ApoB Monoclonal antibody specifically detects Apolipoprotein B/ApoB in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Apolipoprotein B/ApoB
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, Immunoprecipitation
SR1898
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunoprecipitation 1:50
Apo B-100, apoB-100, apoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100
A synthesized peptide derived from human Apolipoprotein B/ApoB (Uniprot #: P04114)
100 μg
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
338
Store at -20° C. Avoid freeze/thaw cycles.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications