Learn More
EAAT3 Antibody (S180-41), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59319-25ug
This item is not returnable.
View return policy
Description
EAAT3 Monoclonal antibody specifically detects EAAT3 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.
Specifications
| EAAT3 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide | |
| SLC1A1 | |
| Fusion protein amino acids 1-524 (full length) of human EAAT3 (Uniprot: P43005) | |
| 25 μg | |
| Primary | |
| Detects 60kDa. | |
| Store at -20°C. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| S180-41 | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| EAAC1excitatory amino acid carrier 1, EAAT3excitatory amino acid transporter 3, Excitatory amino-acid carrier 1, Neuronal and epithelial glutamate transporter, Sodium-dependent glutamate/aspartate transporter 3, solute carrier family 1 (neuronal/epithelial high affinity glutamatetransporter, system Xag), member 1, Solute carrier family 1 member 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6505 | |
| Human, Rat | |
| Purified |