Learn More
Glypican 1 Antibody (S03-2E8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19539-100ul
 View more versions of this product
Product Code. 18613494
  • £500.00 / 100µL
Description
Glypican 1 Monoclonal antibody specifically detects Glypican 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glypican 1
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
2817
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunofluorescence
S03-2E8
Unconjugated
FLJ38078, glypican, glypican 1, glypican proteoglycan 1, glypican-1
A synthetic peptide of human Glypican 1/GPC1 (Uniprot # P35052)