Learn More
IgE Antibody (3H10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11623-0.025mg
This item is not returnable.
View return policy
Description
IgE Monoclonal antibody specifically detects IgE in Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| IgE | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Equine | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 3H10 | |
| Western Blot, ELISA 1:1000 - 1:10000, Immunohistochemistry, Immunohistochemistry-Frozen | |
| IgE, immunoglobulin heavy constant epsilon | |
| Equine IgE | |
| 0.025 mg | |
| Adaptive Immunity, Immunology | |
| 3497 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |