Learn More
NELF-E Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-05978H
This item is not returnable.
View return policy
Description
NELF-E Polyclonal antibody specifically detects NELF-E in Human samples. It is validated for Western Blot
Specifications
| NELF-E | |
| Polyclonal | |
| Western Blot | |
| D6S45, negative elongation factor E, negative elongation factor polypeptide E, NELFE, NELF-ERDmajor histocompatibility complex gene RD, nuclear protein, RD RNA binding protein, RD RNA-binding protein, RDP, RNA-binding protein RD | |
| The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human negative elongation factor polypeptide E using the numbering given in entry NP_002895.3 (GeneID 7936). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 7936 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |