Learn More
Novus Biologicals™ Recombinant Human CMTM2 GST (N-Term) Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
2 μg
Unit Size:
2µg
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Specifications
| For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 146225 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 53.9 kDa |
| Quantity | 2 μg |
| Storage Requirements | Store at -80°C. Avoid freeze/thaw cycles. |
| Gene Alias | chemokine-like factor super family 2, chemokine-like factor superfamily 2, Chemokine-like factor superfamily member 2, CKLF-like MARVEL transmembrane domain containing 2, CKLF-like MARVEL transmembrane domain-containing protein 2, CKLFSF2, FLJ25732, MGC39436 |
| Gene Symbol | CMTM2 |
| Research Category | Cytokine Research |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Show More |