Learn More
ELMOD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-85094
 View more versions of this product
Product Code. 18496150
  • £571.00 / 0.10mL
Description
ELMOD1 Polyclonal antibody specifically detects ELMOD1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELMOD1
Polyclonal
Western Blot 0.04-0.4 ug/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL
DKFZp547C176, ELMO domain containing 1, ELMO domain-containing protein 1, ELMO/CED-12 domain containing 1
This antibody was developed against Recombinant Protein corresponding to amino acids:THFYNIAPEAPTLSHFQQTFCYLMHEFHKFWIEEDPMDIMEFNRVREKFRKRIIKQLQNPDMALCPHFAASEGLINM
0.1 mL
Apoptosis
55531
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunocytochemistry
Unconjugated
PBS (pH 7.2) and 40% Glycerol
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse