Learn More
HCN4 Antibody (S114/10), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-12910MFV500
Product Code. 18437113
  • £508.00 / 0.10mL
Description
HCN4 Monoclonal antibody specifically detects HCN4 in Human, Mouse, Rat, Rabbit samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HCN4
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
Protein G purified
RUO
Primary
Human, Mouse, Rat, Rabbit
Purified
Flow Cytometry
S114/10
Flow Cytometry
hyperpolarization activated cyclic nucleotide-gated cation channel 4, hyperpolarization activated cyclic nucleotide-gated potassium channel 4, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4, SSS2
Fusion protein amino acids 1019-1198 (c-terminus) of rat HCN4
0.1 mL
Neuroscience
10021
Store at 4C in the dark.