Learn More
Thioredoxin-like 5/TRP14/TXNDC17 Antibody (043), DyLight 680 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89879FR
This item is not returnable.
View return policy
Description
Thioredoxin-like 5/TRP14/TXNDC17 Monoclonal antibody specifically detects Thioredoxin-like 5/TRP14/TXNDC17 in Human samples. It is validated for ELISA
Specifications
| Thioredoxin-like 5/TRP14/TXNDC17 | |
| Monoclonal | |
| DyLight 680 | |
| MGC14353, Protein 42-9-9, thioredoxin (Trx)-related protein, 14 kDa, thioredoxin domain containing 17, thioredoxin domain-containing protein 17, thioredoxin-like 5, Thioredoxin-like protein 5, TRP1414 kDa thioredoxin-related protein, TXNL5 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Thioredoxin-like 5/TRP14/TXNDC17 (Q9BRA2; Met1-Asp123). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 043 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 84817 | |
| Store at 4°C in the dark. | |
| IgG |