Learn More
Novus Biologicals™ MLYCD Protein
Product Code. 18413771 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
100μg
Unit Size:
0.50mg
100µg
Product Code. 18413771
Brand: Novus Biologicals™ NBP1989610.5mg
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-305 of HumanSTK16 The Recombinant Human STK16 Protein is derived from E. coli. The Recombinant Human STK16 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. 20 mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 0.1 M NaCl, 1 mM DTT
Molecular Weight (g/mol) 37.2 kDa
Purification Method SDS-PAGE
Quantity 0.5 mg
Source Human
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name MLYCD
Conjugate Unconjugated
Purity or Quality Grade >90%