Learn More
R&D Systems™ Recombinant Human FGF-21 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in MES, Na 2 SO 4 EDTA and DTT with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in MES, Na 2 SO 4 EDTA and DTT.
Unit Size:
25µg
Description
The Recombinant Human FGF-21 Protein is derived from E. coli. The Recombinant Human FGF-21 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in MES, Na 2 SO 4 EDTA and DTT with BSA as a carrier protein. |
| Gene ID (Entrez) | 26291 |
| Molecular Weight (g/mol) | 20.2 kDa |
| Name | FGF-21 |
| Quantity | 25 μg |
| Source | E. coli-derived human FGF-21 protein His29-Ser209,with a N-terminal 5-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |