Learn More
Ly-6G5C Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-83166-0.1ML
Product Code. 18389230
  • £361.00 / 0.10mL
Description
Ly-6G5C Polyclonal specifically detects Ly-6G5C in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ly-6G5C
Polyclonal
PBS, 2% Sucrose
Rabbit
Immunogen affinity purified
RUO
80741
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
lymphocyte antigen 6 complex, locus G5C
The immunogen is a synthetic peptide directed towards the C-terminal region of Human Ly-6G5C
0.1 mL