Learn More
HOMER3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-85056-0.1ML
This item is not returnable.
View return policy
Description
HOMER3 Polyclonal specifically detects HOMER3 in Human samples. It is validated for Western Blot.
Specifications
| HOMER3 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| homer homolog 3 (Drosophila), homer protein homolog 3, Homer, neuronal immediate early gene, 3, Homer-3, VESL3 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of HOMER3 | |
| 0.1 mL | |
| Neuroscience | |
| 9454 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |