Learn More
Uteroglobin/SCGB1A1 Antibody (1F7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals H00007356-M11
Product Code. 18375307
  • £463.00 / 0.01mg
Description
Uteroglobin/SCGB1A1 Monoclonal antibody specifically detects Uteroglobin/SCGB1A1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Uteroglobin/SCGB1A1
Monoclonal
Unconjugated
AAH04481
Mouse
IgG purified
RUO
Primary
Human
Purified
Western Blot, ELISA, Sandwich ELISA
1F7
In 1x PBS, pH 7.4
blastokinin, CC10Urine protein 1, CC16, CCSPCCPBP, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGBUP-1, UP1, Urinary protein 1, uteroglobin
SCGB1A1 (AAH04481, 22 a.a. ~ 91 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EICPSFQRVIETLLMDTPSSYEAAMELFSPDQDMREAGAQLKKLVDTLPQKPRESIIKLMEKIAQSSLCN
0.1 mg
Cytokine Research
7356
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
IgG2a κ