Learn More
HCN1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-04110-100UL
This item is not returnable.
View return policy
Description
HCN1 Polyclonal specifically detects HCN1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| HCN1 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 348980 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| BCNG1, BCNG-1brain cyclic nucleotide gated channel 1, Brain cyclic nucleotide-gated channel 1, HAC-2, hyperpolarization activated cyclic nucleotide-gated potassium channel 1, potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1 | |
| A synthetic peptide of human HCN1 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |